Dimidium bromide ≥95%

Supplier: BeanTown Chemical

225550-1G 225550-250MG
BT225550-1GEA 287.81 USD
BT225550-1G BT225550-250MG
Dimidium bromide ≥95%
Dimidium bromide

CAS: 518-67-2; EC No: 208-256-7; MDL No: MFCD00011757; RTECS: SF7960500 Solid; Molecular Formula: C20H18BrN3; MW: 380.31 Melting Point: 243-248°


  • RTECS: SF7960500
  • Solid

Formula: C₂₀H₁₈BrN₃
Melting Pt: 243…248 °C
MDL Number: MFCD00011757
CAS Number: 518-67-2
EINECS: 208-256-7
UN: 2811
ADR: 6.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e669239c79726ce91f44ef372cbfed426e90f93b&vid=74b03720866852364cb9050de861a4f7c0e5e57b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR