Potassium bromide for FT-IR spectroscopy
Supplier: BeanTown Chemical
225495-100G
225495-25G
BT225495-100GEA
329.14
USD
BT225495-100G
BT225495-25G
Potassium bromide for FT-IR spectroscopy
Potassium bromide
CAS: 7758-02-3; EC No: 231-830-3; MDL No: MFCD00011358; RTECS: TS7650000
Powder; Linear Formula: KBr; MW: 119.00
Melting Point: 734°; Boiling Point: 1435°
Density (g/mL): 2.75; Refractive Index: 1.559
Hygroscopic
- RTECS: TS7650000
- Refractive index: 1.559
- Hygroscopic
- Powder
Formula:
KBr MW: 119 g/mol Boiling Pt: 1435 Melting Pt: 730 Density: 2.75 g/mL |
MDL Number:
MFCD00011358 CAS Number: 7758-02-3 EINECS: 231-830-3 Merck Index: 13,07701 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...