Hydroxylammonium chloride ≥99%
Supplier: BeanTown Chemical
Synonyms:
Hydroxylamine HCl, Hydroxylamine hydrochloride
225430-100G
225430-500G
BT225430-100GEA
31.45
USD
BT225430-100G
BT225430-500G
Hydroxylammonium chloride ≥99%
Hydroxylammonium chloride
CAS: 5470-11-1; EC No: 226-798-2; MDL No: MFCD00051089; RTECS: NC3675000
UN No: UN2923; Haz Class: 8 (6.1); Packing Group: III
Crystalline; Linear Formula: NH2OH·HCl; MW: 69.49
Melting Point: 155-157° (decomposes)
Density (g/mL): 1.67
Hygroscopic
- RTECS: NC3675000
- Hygroscopic
- Crystalline
Formula:
NH₂OH·HCl Melting Pt: 155…157 °C (dec.) Density: 1.67 g/mL |
MDL Number:
MFCD00051089 CAS Number: 5470-11-1 EINECS: 226-798-2 UN: 2923 ADR: 8,III |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...