Dicopper selenide ≥99.5%
Supplier: BeanTown Chemical
Synonyms:
Copper(I) selenide
225410-10G
225410-50G
BT225410-10GEA
86
USD
BT225410-10G
BT225410-50G
Dicopper selenide ≥99.5%
Dicopper selenide
CAS: 20405-64-5; EC No: 243-796-7; MDL No: MFCD00049908
UN No: UN3077; Haz Class: 9; Packing Group: III
2mm & Down Pieces; Molecular Formula: Cu2Se; MW: 206.05
Melting Point: 1113°
Density (g/mL): 6.84
- 2mm & Down Pieces
Formula:
Cu₂Se Melting Pt: 1113 °C Density: 6.84 g/mL |
MDL Number:
MFCD00049908 CAS Number: 20405-64-5 EINECS: 243-796-7 UN: 3077 ADR: 6.1,III |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...