Bromotrispyrrolidinophosphonium hexafluorophosphate ≥97%
Supplier: BeanTown Chemical
Synonyms:
Bromotri(1-pyrrolidinyl)phosphonium hexafluorophosphate, PyBrOP , Bromotripyrrolidinylphosphonium hexafluorophosphate, Bromotri(pyrrolidin-1-yl)phosphonium hexafluorophosphate(V)
225320-1G
225320-5G
BT225320-1GEA
44
USD
BT225320-1G
BT225320-5G
Bromotrispyrrolidinophosphonium hexafluorophosphate ≥97%
Bromo-tris-pyrrolidinophosphonium hexafluorophosphate
CAS: 132705-51-2; MDL No: MFCD00077412
Powder; Molecular Formula: C12H24BrF6N3P2; MW: 466.18
Moisture Sensitive
- Moisture Sensitive
- Powder
Formula:
C₁₂H₂₄BrF₆N₃P₂ |
MDL Number:
MFCD00077412 CAS Number: 132705-51-2 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...