Lecithin (from soy bean)

Supplier: BeanTown Chemical

225315-1KG 225315-250G
BT225315-1KGEA 154.5 USD
BT225315-1KG BT225315-250G
Lecithin (from soy bean)
Lecithin

CAS: 8002-43-5; EC No: 232-307-2; MDL No: MFCD00147406; RTECS: OG7565000 Solid


  • RTECS: OG7565000
  • Solid

MDL Number: MFCD00147406
CAS Number: 8002-43-5
EINECS: 232-307-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR