Bis(triphenylphosphine)palladium(II) chloride ≥99.95% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Bis(triphenylphosphine)palladium(II) dichloride
225100-1G
225100-5G
BT225100-1GEA
66.25
USD
BT225100-1G
BT225100-5G
Bis(triphenylphosphine)palladium(II) chloride ≥99.95% (trace metals basis)
Bis(triphenylphosphine)palladium (II) chloride
CAS: 13965-03-2; EC No: 237-744-2; MDL No: MFCD00009593; RTECS: RT3578000
Crystalline/Powder; Linear Formula: [(C6H5)3P]2PdCl2; Molecular Formula: C36H30Cl2P2Pd; MW: 701.91
Melting Point: 310° (decomposes)
Hygroscopic
- RTECS: RT3578000
- Hygroscopic
- Crystalline/Powder
Formula:
C₃₆H₃₀Cl₂P₂Pd MW: 701.91 g/mol Melting Pt: 310 °C (dec.) |
MDL Number:
MFCD00009593 CAS Number: 13965-03-2 EINECS: 237-744-2 |
Specification Test Results
Assay | 99.95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...