Lactose, anhydrous ≥98%

Supplier: BeanTown Chemical

225065-1KG 225065-500G
BT225065-1KGEA 172.58 USD
BT225065-1KG BT225065-500G
Lactose, anhydrous ≥98%
Lactose

CAS: 63-42-3; EC No: 200-559-2; MDL No: MFCD00151251; RTECS: OD9625000 Powder; Molecular Formula: C12H22O11; MW: 342.30


  • RTECS: OD9625000
  • Powder

Formula: C₁₂H₂₂O₁₁
MDL Number: MFCD00151251
CAS Number: 63-42-3
EINECS: 200-559-2

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR