Cesium nitrate ≥99.99%
Supplier: BeanTown Chemical
Synonyms:
Nitric acid cesium salt
225010-100G
225010-25G
225010-5G
BT225010-100GEA
274.61
USD
BT225010-100G
BT225010-25G
BT225010-5G
Cesium nitrate ≥99.99%
Cesium nitrate
CAS: 7789-18-6; EC No: 232-146-8; MDL No: MFCD00010963; RTECS: FL0700000
UN No: UN1451; Haz Class: 5.1; Packing Group: III
Crystalline; Molecular Formula: CsNO3; MW: 194.91
Melting Point: 414°
Density (g/mL): 3.69
Hygroscopic
- RTECS: FL0700000
- Hygroscopic
- Crystalline
Formula:
CsNO₃ Melting Pt: 414 °C Density: 3.69 g/mL |
MDL Number:
MFCD00010963 CAS Number: 7789-18-6 EINECS: 232-146-8 UN: 1451 ADR: 5.1,III |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...