Silver trifluoromethanesulfonate ≥98%

Supplier: BeanTown Chemical

Synonyms: Silver trifluoromethanesulphonate, Silver triflate, Silver trifluoromethanesulfonate, Trifluoromethanesulphonic acid silver salt, Trifluoromethanesulfonic acid silver salt

225005-25G 225005-5G
BT225005-25GEA 182.57 USD
BT225005-25G BT225005-5G
Silver trifluoromethanesulfonate ≥98%
Silver trifluoromethanesulphonate

CAS: 2923-28-6; EC No: 220-882-2; MDL No: MFCD00013226 UN No: UN3260; Haz Class: 8; Packing Group: III Crystalline; Linear Formula: CF3SO3Ag; MW: 256.93 Melting Point: 356° Light Sensitive


  • Light Sensitive
  • Crystalline

Formula: CAgF₃O₃S
MW: 256.94 g/mol
Melting Pt: 356
MDL Number: MFCD00013226
CAS Number: 2923-28-6
EINECS: 220-882-2
UN: 3260
ADR: 8 III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR