Tungsten(VI) ethoxide
Supplier: BeanTown Chemical
224920-1G
224920-5G
BT224920-1GEA
91
USD
BT224920-1G
BT224920-5G
Tungsten(VI) ethoxide
Tungsten (VI) ethoxide
CAS: 62571-53-3; MDL No: MFCD00156526
UN No: UN3181; Haz Class: 4.1; Packing Group: III
Pieces/Powder; Linear Formula: W(OCH2CH3)6; Molecular Formula: C12H30O6W; MW: 454.21
Boiling Point: 105-110°/0.05 mmHg
Air Sensitive, Moisture Sensitive
- Air Sensitive, Moisture Sensitive
- Pieces/Powder
Formula:
C₁₀H₂₅O₅W Boiling Pt: 105…110 °C/0.05 mmHg |
MDL Number:
MFCD00156526 CAS Number: 62571-53-3 UN: 3181 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...