Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) dichloride
Supplier: BeanTown Chemical
224890-2G
224890-500MG
BT224890-2GEA
550
USD
BT224890-2G
BT224890-500MG
Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) dichloride
Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) dichloride
CAS: 36309-88-3; MDL No: MFCD03095387
Powder; Molecular Formula: C72H48Cl2N6Ru; MW: 1169.20
Light Sensitive
- Light Sensitive
- Powder
Formula:
C₇₂H₄₈Cl₂N₆Ru |
MDL Number:
MFCD03095387 CAS Number: 36309-88-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...