Phenyl bromoacetate ≥98%

Supplier: BeanTown Chemical

Synonyms: Bromoacetic acid Phenyl ester, Phenyl 2-bromoacetate

224775-25G 224775-5G
BT224775-25GEA 121 USD
BT224775-25G BT224775-5G
Phenyl bromoacetate ≥98%
Phenyl bromoacetate

CAS: 620-72-4; MDL No: MFCD00192391 Solid; Linear Formula: BrCH2COOC6H5; Molecular Formula: C8H7BrO2; MW: 215.04 Melting Point: 31-33°; Boiling Point: 134°/15 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.508


  • Solid

Formula: C₈H₇BrO₂
Boiling Pt: 134 °C/15 mmHg
Melting Pt: 31…33 °C
Density: 1.508 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00192391
CAS Number: 620-72-4

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR