Phenyl bromoacetate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Bromoacetic acid Phenyl ester, Phenyl 2-bromoacetate
224775-25G
224775-5G
BT224775-25GEA
121
USD
BT224775-25G
BT224775-5G
Phenyl bromoacetate ≥98%
Phenyl bromoacetate
CAS: 620-72-4; MDL No: MFCD00192391
Solid; Linear Formula: BrCH2COOC6H5; Molecular Formula: C8H7BrO2; MW: 215.04
Melting Point: 31-33°; Boiling Point: 134°/15 mmHg; Flash point:
- Solid
Formula:
C₈H₇BrO₂ Boiling Pt: 134 °C/15 mmHg Melting Pt: 31…33 °C Density: 1.508 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00192391 CAS Number: 620-72-4 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...