3,4-Dimethoxybenzyl alcohol ≥97%

Supplier: BeanTown Chemical

Synonyms: Veratryl alcohol, (3,4-Dimethoxyphenyl)methanol

224690-100G 224690-25G 224690-500G
BT224690-100GEA 109.55 USD
BT224690-100G BT224690-25G BT224690-500G
3,4-Dimethoxybenzyl alcohol ≥97%
3,4-Dimethoxybenzyl alcohol

CAS: 93-03-8; EC No: 202-212-0; MDL No: MFCD00004638 Liquid; Molecular Formula: C9H12O3 ; MW: 168.19 Boiling Point: 296-297°; Flash point: 110°C (230°F) Density (g/mL): 1.157; Refractive Index: 1.552


  • Refractive index: 1.552
  • Liquid

Formula: C₉H₁₂O₃
MW: 168.19 g/mol
Boiling Pt: 296…297 °C
Density: 1.157 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00004638
CAS Number: 93-03-8
EINECS: 202-212-0

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR