4-Hydroxyphenylboronic acid ≥97%
Supplier: BeanTown Chemical
Synonyms:
(4-Hydroxyphenyl)metaboric acid, Dihydroxy(4-hydroxyphenyl)borane, 4-Hydroxybenzeneboronic acid
224610-1G
224610-5G
BT224610-1GEA
51
USD
BT224610-1G
BT224610-5G
4-Hydroxyphenylboronic acid ≥97%
4-Hydroxyphenylboronic acid
CAS: 71597-85-8; MDL No: MFCD01074628
Solid; Molecular Formula: C6H7BO3; MW: 137.93
- Solid
Formula:
C₆H₇BO₃ MW: 137.93 g/mol |
MDL Number:
MFCD01074628 CAS Number: 71597-85-8 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...