(2-Chlorophenyl)acetic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: 2-Chlorophenylacetic acid

224605-10G 224605-250G 224605-50G
BT224605-10GEA 40 USD
BT224605-10G BT224605-250G BT224605-50G
(2-Chlorophenyl)acetic acid ≥98%
(2-Chlorophenyl)acetic acid

CAS: 2444-36-2; EC No: 219-482-0; MDL No: MFCD00004317 Powder; Molecular Formula: C8H7O2Cl; MW: 170.59 Melting Point: 92-95°


  • Powder

Formula: (C₆H₄Cl)CH₂COOH
MW: 170.6 g/mol
Melting Pt: 92…95 °C
MDL Number: MFCD00004317
CAS Number: 2444-36-2
EINECS: 219-482-0

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR