Tetramethoxysilane ≥98%

Supplier: BeanTown Chemical

Synonyms: Tetramethyl orthosilicate

224600-100G 224600-25G 224600-500G
BT224600-100GEA 89.49 USD
BT224600-100G BT224600-25G BT224600-500G
Tetramethoxysilane ≥98%
Tetramethoxysilane

CAS: 681-84-5; EC No: 211-656-4; MDL No: MFCD00008341; RTECS: VV9800000 UN No: UN2606; Haz Class: 6.1 (3); Packing Group: I Liquid; Linear Formula: Si(OCH3)4; Molecular Formula: C4H12O4Si; MW: 152.22 Melting Point: -4°; Boiling Point: 121-122°; Flash point: 26°C (79°F) Density (g/mL): 1.023; Refractive Index: 1.368 Moisture Sensitive


  • RTECS: VV9800000
  • Refractive index: 1.368
  • Moisture Sensitive
  • Liquid

Formula: C₄H₁₂O₄Si
MW: 152.22 g/mol
Boiling Pt: 121
Melting Pt: -2
Density: 1.023 g/mL
Flash Pt: 26 °C (79 °F)
MDL Number: MFCD00008341
CAS Number: 681-84-5
EINECS: 211-656-4
UN: 2606
ADR: 6.1 I

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR