di-Sodium tetraborate decahydrate 99.5-105.0% ACS
Supplier: BeanTown Chemical
Synonyms:
Borax tetra, Sodium tetraborate decahydrate, Sodium Borate decahydrate
224590-500G
BT224590-500GEA
45.08
USD
BT224590-500G
di-Sodium tetraborate decahydrate 99.5-105.0% ACS
di-Sodium tetraborate decahydrate
CAS: 1303-96-4; EC No: 215-540-4; MDL No: MFCD00149193; RTECS: VZ2275000
Crystalline/Powder; Linear Formula: Na2B4O7·10H2O; MW: 381.37
Melting Point: 62°
Density (g/mL): 1.73
- RTECS: VZ2275000
- Crystalline/Powder
Formula:
Na₂B₄O₇·10H₂O Melting Pt: 62 °C Density: 1.73 g/mL |
MDL Number:
MFCD00149193 CAS Number: 1303-96-4 EINECS: 215-540-4 |
Specification Test Results
Assay | 99.5-105.0% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...