Ethyl-2-chloro-4,4,4-trifluoroacetoacetate ≥94%
Supplier: BeanTown Chemical
Synonyms:
Ethyl 2-chloro-4,4,4-trifluoro-3-oxobutanoate
224530-25G
224530-5G
BT224530-25GEA
180
USD
BT224530-25G
BT224530-5G
Ethyl-2-chloro-4,4,4-trifluoroacetoacetate ≥94%
Ethyl-2-chloro-4,4,4-trifluoroacetoacetate
CAS: 363-58-6; MDL No: MFCD00041540
UN No: UN1760; Haz Class: 8; Packing Group: III
Liquid; Molecular Formula: C6H6ClF3O3; MW: 218.56
Boiling Point: 67-71°/35 mmHg; Flash point: 67°C (153°F)
Density (g/mL): 1.39
- Liquid
Formula:
C₆H₆ClF₃O₃ Boiling Pt: 67…71 °C/35 mmHg Density: 1.39 g/mL Flash Pt: 67 °C (153 °F) |
MDL Number:
MFCD00041540 CAS Number: 363-58-6 ADR: 8,III |
Specification Test Results
Assay | 94% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...