Aluminium ≥99.5% (trace metals basis), powder -100+325 mesh

Supplier: BeanTown Chemical

Synonyms: Aluminum

224515-1KG 224515-250G 224515-50G
BT224515-1KGEA 213 USD
BT224515-1KG BT224515-250G BT224515-50G
Aluminium ≥99.5% (trace metals basis), powder -100+325 mesh
Aluminium

CAS: 7429-90-5; EC No: 231-072-3; MDL No: MFCD00134029; RTECS: BD0330000 UN No: UN1396; Haz Class: 4.3; Packing Group: II Powder; Molecular Formula: Al; MW: 26.98 Melting Point: 660.4°; Boiling Point: 2460° Density (g/mL): 2.7


  • RTECS: BD0330000
  • Powder

Formula: Al
MW: 26.98 g/mol
Boiling Pt: 2327
Melting Pt: 660
Density: 2.7 g/mL
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
UN: 1396
ADR: 4.3 II
Merck Index: 13,00321

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR