(S)-1,2,3,4-Tetrahydroisoquinoline-3-methanol ≥98%
Supplier: BeanTown Chemical
Synonyms:
(S)-(-)-1,2,3,4-Tetrahydro-3-isoquinolinemethanol, (S)-(1,2,3,4-Tetrahydroisoquinolin-3-yl)methanol, (S)-3-Hydroxymethyl-1,2,3,4-tetrahydroisoquinoline
224425-1G
BT224425-1GEA
150
USD
BT224425-1G
(S)-1,2,3,4-Tetrahydroisoquinoline-3-methanol ≥98%
(S)-1,2,3,4-Tetrahydroisoquinoline-3-methanol
CAS: 18881-17-9; MDL No: MFCD01631316
Powder; Molecular Formula: C10H13NO; MW: 163.22
Melting Point: 114-116°
Optical Rotation: [α]22/D -97°, c = 1 in methanol
- Optical rotation: [α]22/D -97°, c = 1 in methanol
- Powder
Formula:
C₁₀H₁₃NO Melting Pt: 114…116 °C |
MDL Number:
MFCD01631316 CAS Number: 18881-17-9 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...