4-Aminobutyric acid ≥97%
Supplier: BeanTown Chemical
Synonyms:
GABA, γ-Aminobutyric acid
224415-100G
224415-500G
BT224415-100GEA
47.07
USD
BT224415-100G
BT224415-500G
4-Aminobutyric acid ≥97%
4-Aminobutyric acid
CAS: 56-12-2; EC No: 200-258-6; MDL No: MFCD00008226; RTECS: ES6300000
Crystalline; Molecular Formula: C4H9NO2; MW: 103.12
Melting Point: 195° (decomposes)
- RTECS: ES6300000
- Crystalline
Formula:
C₄H₉NO₂ Melting Pt: 195 °C (dec.) |
MDL Number:
MFCD00008226 CAS Number: 56-12-2 EINECS: 200-258-6 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...