Magnesium chloride hexahydrate 99.0-102.0% ACS

Supplier: BeanTown Chemical

Synonyms: Magnesium dichloride

224315-2KG 224315-500G
BT224315-2KGEA 146.73 USD
BT224315-2KG BT224315-500G
Magnesium chloride hexahydrate 99.0-102.0% ACS
Magnesium chloride hexahydrate

CAS: 7791-18-6; EC No: 232-094-6; MDL No: MFCD00149781; RTECS: OM2975000 Crystalline; Linear Formula: MgCl2·6H2O; MW: 203.30 Melting Point: 117° (decomposes) Density (g/mL): 1.57 Hygroscopic, Moisture Sensitive


  • RTECS: OM2975000
  • Hygroscopic, Moisture Sensitive
  • Crystalline

Formula: MgCl₂·6H₂O
MW: 203.3 g/mol
Boiling Pt: 1412
Melting Pt: 117 (dec.)
Density: 1.57 g/mL
MDL Number: MFCD00149781
CAS Number: 7791-18-6
EINECS: 232-094-6
Merck Index: 13,05684

Order Now

Specification Test Results

Assay 99.0-102.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR