Gadolinium ≥99.9% (trace rare earth metals basis), foil, Thickness 0.127 mm

Supplier: BeanTown Chemical

224280-25X25MM 224280-50X50MM
BT224280-25X25EA 90 USD
BT224280-25X25 BT224280-50X50
Gadolinium ≥99.9% (trace rare earth metals basis), foil, Thickness 0.127 mm
Gadolinium

CAS: 7440-54-2; EC No: 231-162-2; MDL No: MFCD00011022; RTECS: LW3850000 UN No: UN3178; Haz Class: 4.1; Packing Group: II Solid; Molecular Formula: Gd; MW: 157.25 Melting Point: 1313°; Boiling Point: 3273° Density (g/mL): 7.886


  • RTECS: LW3850000
  • Solid

Formula: Gd
MW: 157.25
Boiling Pt: 3273 °C
Melting Pt: 1313 °C
Density: 7.886 g/mL
MDL Number: MFCD00011022
CAS Number: 7440-54-2
EINECS: 231-162-2
ADR: 4.1 II

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR