4'-Bromoacetophenone ≥98%
Supplier: BeanTown Chemical
Synonyms:
p-Bromoacetophenone, 1-(4-Bromophenyl)ethanone
224250-100G
224250-25G
224250-500G
BT224250-100GEA
73.4
USD
BT224250-100G
BT224250-25G
BT224250-500G
4'-Bromoacetophenone ≥98%
4'-Bromoacetophenone
CAS: 99-90-1; EC No: 202-799-3; MDL No: MFCD00000105
Solid; Molecular Formula: C8H7BrO; MW: 199.04
Melting Point: 49-51°; Boiling Point: 255°; Flash point:
- Solid
Formula:
C₈H₇BrO Boiling Pt: 255 °C Melting Pt: 49…51 °C Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00000105 CAS Number: 99-90-1 EINECS: 202-799-3 UN: 2811 ADR: 6.1,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...