Platinum iridium alloy (80:20 w%) ≥99.9% (trace metals basis), foil, Thickness 0.1 mm

Supplier: BeanTown Chemical

224240-25X25MM 224240-50X50MM
BT224240-25X25EA 354.47 USD
BT224240-25X25 BT224240-50X50
Platinum iridium alloy (80:20 w%) ≥99.9% (trace metals basis), foil, Thickness 0.1 mm
Platinum Iridium alloy (80:20 w%)

MDL No: MFCD02091683 Solid; Formula: Pt:Ir; 80:20 wt%


  • Solid

MDL Number: MFCD02091683
UN: 3178
ADR: 4.1,III

Order Now

Specification Test Results

Assay 99.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR