N,N-Dimethylaminophosphoryl dichloride ≥97%

Supplier: BeanTown Chemical

Synonyms: N,N-Dimethylphosphoramidodichloridate

224235-25G 224235-5G
BT224235-25GEA 160 USD
BT224235-25G BT224235-5G
N,N-Dimethylaminophosphoryl dichloride ≥97%
N,N-Dimethylaminophosphoryl dichloride

CAS: 677-43-0; EC No: 211-641-2; MDL No: MFCD00013896; RTECS: TB4025000 UN No: UN3265; Haz Class: 8; Packing Group: II Liquid; Linear Formula: (CH3)2NP(O)Cl2; Molecular Formula: C2H6Cl2NOP; MW: 161.96 Boiling Point: 77-79°/11 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.362; Refractive Index: 1.464 Air Sensitive, Hygroscopic, Moisture Sensitive


  • RTECS: TB4025000
  • Refractive index: 1.464
  • Air Sensitive, Hygroscopic, Moisture Sensitive
  • Liquid

Formula: C₂H₆Cl₂NOP
MW: 161.95 g/mol
Boiling Pt: 77…79 °C/11 mmHg
Density: 1.362 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00013896
CAS Number: 677-43-0
EINECS: 211-641-2
UN: 3265
ADR: 8 II

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR