Pyridoxine hydrochloride ≥99%
Supplier: BeanTown Chemical
Synonyms:
Adermine hydrochloride, Vitamin B6 hydrochloride, PN HCl, Pyridoxine HCl, (5-Hydroxy-6-methylpyridine-3,4-diyl)dimethanol hydrochloride, Pyridoxol hydrochloride, Pyridoxine HCI
224220-100G
224220-25G
224220-500G
BT224220-100GEA
100
USD
BT224220-100G
BT224220-25G
BT224220-500G
Pyridoxine hydrochloride ≥99%
Pyridoxine hydrochloride
CAS: 58-56-0; EC No: 200-386-2; MDL No: MFCD00012807; RTECS: UV1350000
Crystalline/Powder; Molecular Formula: C8H11NO3·HCl; MW: 205.64
Melting Point: 214-215°
Light Sensitive
- RTECS: UV1350000
- Light Sensitive
- Crystalline/Powder
Formula:
C₈H₁₂ClNO₃ Melting Pt: 214…215 °C |
MDL Number:
MFCD00012807 CAS Number: 58-56-0 EINECS: 200-386-2 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...