Zincon sodium salt

Supplier: BeanTown Chemical

224205-1G 224205-25G 224205-5G
BT224205-1GEA 31.63 USD
BT224205-1G BT224205-25G BT224205-5G
Zincon sodium salt
Zincon sodium salt

CAS: 62625-22-3; EC No: 263-651-1; MDL No: MFCD00064385 Solid; Molecular Formula: C20H15N4NaO6S; MW: 462.42 Melting Point: 203° (decomposes)


  • Solid

Formula: C₂₀H₁₅N₄NaO₆S
MW: 462.412 g/mol
Melting Pt: 203 °C (dec.)
MDL Number: MFCD00064385
CAS Number: 62625-22-3
EINECS: 263-651-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR