4-(2-Hydroxyethyl)benzonitrile ≥98%

Supplier: BeanTown Chemical

224190-1G 224190-250MG 224190-5G
BT224190-1GEA 223 USD
BT224190-1G BT224190-250MG BT224190-5G
4-(2-Hydroxyethyl)benzonitrile ≥98%
4-(2-Hydroxyethyl)benzonitrile

CAS: 69395-13-7; MDL No: MFCD00052461 UN No: UN3439; Haz Class: 6.1; Packing Group: III Powder/Chunks; Molecular Formula: C9H9NO; MW: 147.18


  • Powder/Chunks

Formula: C₉H₉NO
MW: 147.18 g/mol
MDL Number: MFCD00052461
CAS Number: 69395-13-7
ADR: 6.1 III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR