Gold ≥99.995% (trace metals basis), wire Ø 0.05 mm
Supplier: BeanTown Chemical
224110-100M
224110-25M
224110-5M
BT224110-100MEA
1222
USD
BT224110-100M
BT224110-25M
BT224110-5M
Gold ≥99.995% (trace metals basis), wire Ø 0.05 mm
Gold
CAS: 7440-57-5; EC No: 231-165-9; MDL No: MFCD00003436; RTECS: MD5420000
Solid; Molecular Formula: Au; MW: 196.97
Melting Point: 1063°; Boiling Point: 2808°
Density (g/mL): 19.3
- RTECS: MD5420000
- Solid
Formula:
Au Boiling Pt: 2808 °C Melting Pt: 1063 °C Density: 19.3 g/mL |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 |
Specification Test Results
Assay | 99.995% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...