Ruthenium(IV) oxide, anhydrous ≥99.95%

Supplier: BeanTown Chemical

Synonyms: Ruthenium dioxide, Ruthenium tetroxide

224080-10G 224080-2G 224080-500MG
BT224080-10GEA 858 USD
BT224080-10G BT224080-2G BT224080-500MG
Ruthenium(IV) oxide, anhydrous ≥99.95%
Ruthenium (IV) oxide

CAS: 12036-10-1; EC No: 234-840-6; MDL No: MFCD00011210; RTECS: VM2654000 UN No: UN1479; Haz Class: 5.1; Packing Group: II Sub-micron Powder; Linear Formula: RuO2; MW: 133.07 Melting Point: 1200° (sublimes) Density (g/mL): 6.97 Hygroscopic


  • RTECS: VM2654000
  • Hygroscopic
  • Sub-micron Powder

Formula: O₂Ru
MW: 133.07 g/mol
Melting Pt: 1200 (subl.)
Density: 6.97 g/mL
MDL Number: MFCD00074857
CAS Number: 12036-10-1
EINECS: 234-840-6
UN: 1479
ADR: 5.1 II

Order Now

Specification Test Results

Assay 99.95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR