(S)-(+)-2-Amino-1-phenylethanol ≥97%
Supplier: BeanTown Chemical
224000-1G
224000-5G
BT224000-1GEA
75
USD
BT224000-1G
BT224000-5G
(S)-(+)-2-Amino-1-phenylethanol ≥97%
(S)-(+)-2-Amino-1-phenylethanol
CAS: 56613-81-1; MDL No: MFCD00239405
UN No: UN3259; Haz Class: 8; Packing Group: III
Powder; Molecular Formula: C8H11NO ; MW: 137.18
Melting Point: 57-59°; Boiling Point: 160°/17 mmHg
Optical Rotation: [α]20/D +43±2°, c = 2% in ethanol
Air Sensitive
- Optical rotation: [α]20/D +43±2°, c = 2% in ethanol
- Air Sensitive
- Powder
Formula:
C₈H₁₁NO Boiling Pt: 160 °C/17 mmHg Melting Pt: 57…59 °C |
MDL Number:
MFCD00239405 CAS Number: 56613-81-1 ADR: 8,III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...