2,3,6-Trifluorophenol ≥98%
Supplier: BeanTown Chemical
223980-1G
223980-5G
BT223980-1GEA
34
USD
BT223980-1G
BT223980-5G
2,3,6-Trifluorophenol ≥98%
2,3,6-Trifluorophenol
CAS: 113798-74-6; MDL No: MFCD00061214
UN No: UN3261; Haz Class: 8; Packing Group: II
Liquid/Low Melting Solid; Molecular Formula: C6H3F3O; MW: 148.08
Melting Point: 33-37°; Flash point: 64°C (147°F)
- Liquid/Low Melting Solid
Formula:
C₆H₃F₃O Melting Pt: 33…37 °C Flash Pt: 64 °C (147 °F) |
MDL Number:
MFCD00061214 CAS Number: 113798-74-6 UN: 3261 ADR: 8,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...