L(-)-Carnitine hydrochloride
Supplier: BeanTown Chemical
Synonyms:
Levocarnitine hydrochloride, L-Carnitine hydrochloride, (R)-3-Carboxy-2-hydroxy-N,N,N-trimethylpropan-1-aminium chloride, LC-80, (R)-3-Hydroxy-4-(trimethylammonio)butyrate hydrochloride, L-Carnitine chloride, (R)-Carnitine hydrochloride, Levocarnitine chloride
223975-1G
BT223975-1GEA
479
USD
BT223975-1G
L(-)-Carnitine hydrochloride
L(-)-Carnitine hydrochloride
CAS: 10017-44-4; EC No: 229-663-6; MDL No: MFCD00066100; RTECS: BP2979100
Solid; Linear Formula: (CH3)3NCH2CH(OH)COOH·Cl; Molecular Formula: C7H16ClNO3; MW: 197.66
Melting Point: 142°
- RTECS: BP2979100
- Solid
Formula:
C₇H₁₆ClNO₃ Melting Pt: 142 °C |
MDL Number:
MFCD00066100 CAS Number: 6645-46-1 EINECS: 229-663-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...