D-(-)-N-Ethylglucamine ≥98%
Supplier: BeanTown Chemical
Synonyms:
N-Ethyl-D-glucamine, (-)-1-Deoxy-1-(ethylamino)-D-glucitol, (-)-1-Ethylamino-1-deoxy-D-glucitol, (2R,3R,4R,5S)-6-(Ethylamino)hexane-1,2,3,4,5-pentaol, (-)-N-Ethyl-D-glucamine
223955-100G
BT223955-100GEA
44
USD
BT223955-100G
D-(-)-N-Ethylglucamine ≥98%
D(-)-N-Ethylglucamine
CAS: 14216-22-9; EC No: 238-073-8; MDL No: MFCD03789564
Powder; Molecular Formula: C8H19NO5; MW: 209.24
Melting Point: 136-140°
Optical Rotation: [α]/D -17±2.0°, c = 1% in water
Air Sensitive, Hygroscopic
- Optical rotation: [α]/D -17±2.0°, c = 1% in water
- Air Sensitive, Hygroscopic
- Powder
Formula:
H₃CCH₂NHCH₂(CH(OH))₄CH₂OH Melting Pt: 136…140 °C |
MDL Number:
MFCD03789564 CAS Number: 14216-22-9 EINECS: 238-073-8 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...