9,10-Bis(phenylethynyl)anthracene

Supplier: BeanTown Chemical

223835-1G 223835-25G 223835-5G
BT223835-1GEA 35.17 USD
BT223835-1G BT223835-25G BT223835-5G
9,10-Bis(phenylethynyl)anthracene
9,10-Bis(phenylethynyl)anthracene

CAS: 10075-85-1; EC No: 233-210-8; MDL No: MFCD00012050 Crystalline; Molecular Formula: C30H18; MW: 378.46 Melting Point: 248-250° Light Sensitive


  • Light Sensitive
  • Crystalline

Formula: C₃₀H₁₈
Melting Pt: 248…250 °C
MDL Number: MFCD00012050
CAS Number: 10075-85-1
EINECS: 233-210-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR