Phthalaldehyde ≥98%

Supplier: BeanTown Chemical

Synonyms: 1,2-Phthalic dicarboxaldehyde, Phthaldialdehyde, o-Phthalaldehyde, OPA, Phthaldicarboxaldehyde, Benzene-1,2-dicarboxaldehyde

223690-10G 223690-250G 223690-50G
BT223690-10GEA 77.44 USD
BT223690-10G BT223690-250G BT223690-50G
Phthalaldehyde ≥98%
Phthalaldehyde

CAS: 643-79-8; EC No: 211-402-2; MDL No: MFCD00003335; RTECS: TH6950000 UN No: UN2923; Haz Class: 8 (6.1); Packing Group: II Powder; Molecular Formula: C8H6O2; MW: 134.14 Melting Point: 55-58°; Boiling Point: 83°/0.800 mmHg; Flash point: 132°C (269°F) Light Sensitive, Moisture Sensitive


  • RTECS: TH6950000
  • Light Sensitive, Moisture Sensitive
  • Powder

Formula: C₈H₆O₂
MW: 134.13 g/mol
Boiling Pt: 266
Melting Pt: 55…58 °C
Flash Pt: 132 °C (269 °F)
MDL Number: MFCD00003335
CAS Number: 643-79-8
EINECS: 211-402-2
UN: 2923
ADR: 8 II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR