Zirconium(IV) oxide ≥99% stabilised

Supplier: BeanTown Chemical

Synonyms: Zirconium dioxide, Zirconium Oxide

Stabilization: Stabilised with yttria (Y₂O₃)

223670-100G 223670-500G
BT223670-100GEA 95 USD
BT223670-100G BT223670-500G
Zirconium(IV) oxide ≥99% stabilised
Zirconium (IV) oxide

CAS: 1314-23-4; EC No: 215-227-2; MDL No: MFCD00145546; RTECS: ZH8800000 50nm Powder; Linear Formula: ZrO2 + 10-15% Y2O3; MW: Melting Point: 2700°; Boiling Point: 5000°


  • RTECS: ZH8800000
  • <50nm Powder

Formula: ZrO₂
Boiling Pt: 5000 °C
Melting Pt: 2700 °C
MDL Number: MFCD00011310
CAS Number: 1314-23-4
EINECS: 215-227-2

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR