Sodium benzoate ≥99%

Supplier: BeanTown Chemical

Synonyms: Benzoic acid sodium salt

223645-2.5KG 223645-500G
BT223645-2.5KGEA 83.58 USD
BT223645-2.5KG BT223645-500G
Sodium benzoate ≥99%
Sodium benzoate

CAS: 532-32-1; EC No: 208-534-8; MDL No: MFCD00012463; RTECS: DH6650000 Powder; Linear Formula: C6H5COONa; Molecular Formula: C7H5NaO2; MW: 144.11 Melting Point: 300°; Flash point: 100°C (212°F) Density (g/mL): 1.44 Hygroscopic


  • RTECS: DH6650000
  • Hygroscopic
  • Powder

Formula: C₆H₅COONa
Melting Pt: >300 °C
Density: 1.44 g/mL
Flash Pt: >100 °C (>212 °F)
MDL Number: MFCD00012463
CAS Number: 532-32-1
EINECS: 208-534-8

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR