Potassium hydrogen phthalate 99.95-100.05% ACS primary standard
Supplier: BeanTown Chemical
Synonyms:
Potassium phthalate monobasic, Potassium biphthalate, Potassium Acid Phthalate, KHP, Phthalic Acid Monopotassium Salt, Phthalic acid potassium salt
223510-1KG
223510-250G
BT223510-1KGEA
148.36
USD
BT223510-1KG
BT223510-250G
Potassium hydrogen phthalate 99.95-100.05% ACS primary standard
Potassium hydrogen phthalate
CAS: 877-24-7; EC No: 212-889-4; MDL No: MFCD00013070; RTECS: CZ4326000
Crystalline/Powder; Linear Formula: HOOCC6H4COOK; Molecular Formula: C8H5KO4; MW: 204.23
Melting Point: 295-300° (decomposes)
Density (g/mL): 1.636
- RTECS: CZ4326000
- Crystalline/Powder
Formula:
HOOCC₆H₄COOK MW: 204.22 g/mol Melting Pt: 295…300 °C (dec.) Density: 1.636 g/mL |
MDL Number:
MFCD00013070 CAS Number: 877-24-7 EINECS: 212-889-4 Merck Index: 13,07694 |
|
Specification Test Results
Assay | 99.95-100.05% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...