Bismuth ≥99.997%, granules 1-2 mm
Supplier: BeanTown Chemical
223485-100G
223485-2KG
223485-500G
BT223485-100GEA
143.98
USD
BT223485-100G
BT223485-2KG
BT223485-500G
Bismuth ≥99.997%, granules 1-2 mm
Bismuth
CAS: 7440-69-9; EC No: 231-177-4; MDL No: MFCD00134033; RTECS: EB2600000
Solid; Molecular Formula: Bi; MW: 208.98
Melting Point: 271°; Boiling Point: 1560°
Density (g/mL): 9.8
- RTECS: EB2600000
- Solid
Formula:
Bi Boiling Pt: 1560 °C Melting Pt: 271 °C Density: 9.8 g/mL |
MDL Number:
MFCD00134033 CAS Number: 7440-69-9 EINECS: 231-177-4 |
Specification Test Results
Assay | 99.997% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...