Bismuth ≥99.997%, granules 1-2 mm

Supplier: BeanTown Chemical

223485-100G 223485-2KG 223485-500G
BT223485-100GEA 143.98 USD
BT223485-100G BT223485-2KG BT223485-500G
Bismuth ≥99.997%, granules 1-2 mm
Bismuth

CAS: 7440-69-9; EC No: 231-177-4; MDL No: MFCD00134033; RTECS: EB2600000 Solid; Molecular Formula: Bi; MW: 208.98 Melting Point: 271°; Boiling Point: 1560° Density (g/mL): 9.8


  • RTECS: EB2600000
  • Solid

Formula: Bi
Boiling Pt: 1560 °C
Melting Pt: 271 °C
Density: 9.8 g/mL
MDL Number: MFCD00134033
CAS Number: 7440-69-9
EINECS: 231-177-4

Order Now

Specification Test Results

Assay 99.997%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR