Platinium(IV) oxide, anhydrous ≥99.95%

Supplier: BeanTown Chemical

Synonyms: Platinum dioxide, Adam's type catalyst

223435-1G 223435-250MG 223435-5G
BT223435-1GEA 255.67 USD
BT223435-1G BT223435-250MG BT223435-5G
Platinium(IV) oxide, anhydrous ≥99.95%
Platinium (IV) oxide

CAS: 1314-15-4; EC No: 215-223-0; MDL No: MFCD00011184; RTECS: TP2506020 UN No: UN1479; Haz Class: 5.1; Packing Group: II Powder; Linear Formula: PtO2; MW: 227.09 Melting Point: 450°


  • RTECS: TP2506020
  • Powder

Formula: O₂Pt
MW: 227.08
Melting Pt: 450
Density: 11.8
MDL Number: MFCD00011184
CAS Number: 1314-15-4
EINECS: 215-223-0
UN: 1479
ADR: 5.1 II
Merck Index: 12,07686

Order Now

Specification Test Results

Assay 99.95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR