Mercury(I) nitrate dihydrate ≥98%

Supplier: BeanTown Chemical

Synonyms: Dimercury dinitrate dihydrate, Mercurous nitrate dihydrate

223415-1KG 223415-250G 223415-50G
BT223415-1KGEA 599 USD
BT223415-1KG BT223415-250G BT223415-50G
Mercury(I) nitrate dihydrate ≥98%
Mercury (I) nitrate dihydrate

CAS: 14836-60-3; EC No: 233-886-4; MDL No: MFCD00150812; RTECS: OW8000000 UN No: UN1627; Haz Class: 6.1; Packing Group: II Crystalline; Linear Formula: Hg2(NO3)2·2H2O; MW: 561.22 Melting Point: 70° Density (g/mL): 4.78 Light Sensitive


  • RTECS: OW8000000
  • Light Sensitive
  • Crystalline

Formula: Hg₂N₂O₆·2H₂O
MW: 561.22 g/mol
Melting Pt: 70
Density: 4.78 g/mL
MDL Number: MFCD00150812
CAS Number: 14836-60-3
EINECS: 233-886-4
UN: 1627
ADR: 6.1 II
Merck Index: 13,05923

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR