Titanium(IV) isopropoxide ≥97%
Supplier: BeanTown Chemical
Synonyms:
Titanium tetraisopropanolate, Titanium (IV) isopropoxide
223400-100G
223400-500G
BT223400-100GEA
53.77
USD
BT223400-100G
BT223400-500G
Titanium(IV) isopropoxide ≥97%
Titanium (IV) isopropoxide
CAS: 546-68-9; EC No: 208-909-6; MDL No: MFCD00008871; RTECS: NT8060000
UN No: UN2413; Haz Class: 3; Packing Group: III
Liquid; Linear Formula: Ti[OCH(CH3)2]4; Molecular Formula: C12H28O4Ti ; MW: 284.23
Melting Point: 14-17°; Boiling Point: 232°; Flash point: 45°C (113°F)
Density (g/mL): 0.955; Refractive Index: 1.464
Moisture Sensitive
- RTECS: NT8060000
- Refractive index: 1.464
- Moisture Sensitive
- Liquid
Formula:
C₁₂H₂₈O₄Ti MW: 284.23 g/mol Boiling Pt: 240 Melting Pt: 14…17 °C Density: 0.955 g/mL Flash Pt: 45 °C (113 °F) |
MDL Number:
MFCD00008871 CAS Number: 546-68-9 EINECS: 208-909-6 UN: 2413 ADR: 3 III Merck Index: 12,09614 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...