Gadolinium ≥99.9% (trace metals basis excluding Ta), rods Ø 6.35 mm

Supplier: BeanTown Chemical

223380-10MM 223380-25MM 223380-50MM
BT223380-10MMEA 448 USD
BT223380-10MM BT223380-25MM BT223380-50MM
Gadolinium ≥99.9% (trace metals basis excluding Ta), rods Ø 6.35 mm
Gadolinium

CAS: 7440-54-2; EC No: 231-162-2; MDL No: MFCD00011022 Solid, cast rod, diameter is approximate; Molecular Formula: Gd; MW: 157.25 Melting Point: 1313°; Boiling Point: 3273° Density (g/mL): 7.886


  • Solid, cast rod, diameter is approximate

Formula: Gd
MW: 157.25
Boiling Pt: 3273 °C
Melting Pt: 1313 °C
Density: 7.886 g/mL
MDL Number: MFCD00011022
CAS Number: 7440-54-2
EINECS: 231-162-2

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR