Ammonium thiocyanate ≥98%
Supplier: BeanTown Chemical
223345-1KG
223345-250G
223345-5KG
BT223345-1KGEA
170.46
USD
BT223345-1KG
BT223345-250G
BT223345-5KG
Ammonium thiocyanate ≥98%
Ammonium thiocyanate
CAS: 1762-95-4; EC No: 217-175-6; MDL No: MFCD00011428; RTECS: XK7875000
Powder; Linear Formula: NH4SCN; MW: 76.12
Melting Point: 152-154°
Hygroscopic , Light Sensitive
- RTECS: XK7875000
- Hygroscopic, Light Sensitive
- Powder
Formula:
NH₄SCN Melting Pt: 152…154 °C |
MDL Number:
MFCD00011428 CAS Number: 1762-95-4 EINECS: 217-175-6 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...