Manganese(II) chloride tetrahydrate ≥99.999%

Supplier: BeanTown Chemical

Synonyms: Manganese dichloride tetrahydrate, Manganese chloride (tetrahydrate), Manganese(II) chloride tetrahydrate, Manganous chloride tetrahydrate, Manganese chloride tetrahydrate, Manganese Chloride, Tetrahydrate

223325-10G 223325-50G
BT223325-10GEA 80 USD
BT223325-10G BT223325-50G
Manganese(II) chloride tetrahydrate ≥99.999%
Manganese (II) chloride tetrahydrate

CAS: 13446-34-9; EC No: 231-869-6; MDL No: MFCD00149792 Crystalline; Linear Formula: MnCl2·4H2O; MW: 197.90 Melting Point: 58° Density (g/mL): 1.913 Hygroscopic


  • Hygroscopic
  • Crystalline

Formula: MnCl₂·4H₂O
Melting Pt: 58 °C
Density: 1.913 g/mL
MDL Number: MFCD00149792
CAS Number: 13446-34-9
EINECS: 231-869-6
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Assay 99.999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR